3-{[(2,4-Difluorobenzyl)oxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

050276-500MG 050276-2.5G
101891-004EA 364.62 USD
101891-004 101891-006
3-{[(2,4-Difluorobenzyl)oxy]methyl}pyrrolidine hydrochloride
3-{[(2,4-Difluorobenzyl)oxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆ClF₂NO
MDL Number: MFCD13560677
CAS Number: 1219976-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR