2-{2-[(2,4-Difluorobenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050275-500MG 050275-2.5G
101891-000EA 364.62 USD
101891-000 101891-002
2-{2-[(2,4-Difluorobenzyl)oxy]ethyl}piperidine hydrochloride
2-{2-[(2,4-Difluorobenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₀ClF₂NO
MDL Number: MFCD13560676
CAS Number: 1220016-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR