3-{2-[(2,4-Difluorobenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050274-500MG 050274-2.5G
101890-996EA 364.62 USD
101890-996 101890-998
3-{2-[(2,4-Difluorobenzyl)oxy]ethyl}piperidine hydrochloride
3-{2-[(2,4-Difluorobenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₀ClF₂NO
MDL Number: MFCD13560675
CAS Number: 1220032-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR