4-Iodobenzyl-3-pyrrolidinylmethyl ether hydrochloride
Supplier: Matrix Scientific
050272-500MG
050272-2.5G
101890-988EA
364.62
USD
101890-988
101890-990
4-Iodobenzyl-3-pyrrolidinylmethyl ether hydrochloride
4-Iodobenzyl-3-pyrrolidinylmethyl ether hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₇ClINO |
MDL Number:
MFCD13560673 CAS Number: 1220016-42-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...