3-{[(4-Bromobenzyl)oxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

050268-500MG 050268-2.5G
101890-972EA 364.62 USD
101890-972 101890-974
3-{[(4-Bromobenzyl)oxy]methyl}pyrrolidine hydrochloride
3-{[(4-Bromobenzyl)oxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇BrClNO
MDL Number: MFCD13560669
CAS Number: 1219964-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR