2-{2-[(4-Bromobenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050267-500MG 050267-2.5G
101890-968EA 364.62 USD
101890-968 101890-970
2-{2-[(4-Bromobenzyl)oxy]ethyl}piperidine hydrochloride
2-{2-[(4-Bromobenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁BrClNO
MDL Number: MFCD13560668
CAS Number: 1220032-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR