3-{2-[(4-Bromobenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050266-500MG 050266-2.5G
101890-964EA 364.62 USD
101890-964 101890-966
3-{2-[(4-Bromobenzyl)oxy]ethyl}piperidine hydrochloride
3-{2-[(4-Bromobenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁BrClNO
MDL Number: MFCD13560667
CAS Number: 1220038-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR