4-{2-[(4-Bromobenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050265-500MG 050265-2.5G
101890-960EA 364.62 USD
101890-960 101890-962
4-{2-[(4-Bromobenzyl)oxy]ethyl}piperidine hydrochloride
4-{2-[(4-Bromobenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁BrClNO
MDL Number: MFCD13560666
CAS Number: 1220016-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR