3-{[(3-Bromobenzyl)oxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

050264-500MG 050264-2.5G
101890-956EA 364.62 USD
101890-956 101890-958
3-{[(3-Bromobenzyl)oxy]methyl}pyrrolidine hydrochloride
3-{[(3-Bromobenzyl)oxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇BrClNO
MDL Number: MFCD13560665
CAS Number: 1220038-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR