2-{2-[(3-Bromobenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050263-500MG 050263-2.5G
101890-952EA 364.62 USD
101890-952 101890-954
2-{2-[(3-Bromobenzyl)oxy]ethyl}piperidine hydrochloride
2-{2-[(3-Bromobenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁BrClNO
MDL Number: MFCD13560664
CAS Number: 1220016-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR