3-{2-[(3-Bromobenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050262-500MG 050262-2.5G
101890-948EA 364.62 USD
101890-948 101890-950
3-{2-[(3-Bromobenzyl)oxy]ethyl}piperidine hydrochloride
3-{2-[(3-Bromobenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁BrClNO
MDL Number: MFCD13560663
CAS Number: 1220032-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR