4-{2-[(3-Bromobenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050261-500MG 050261-2.5G
101890-944EA 364.62 USD
101890-944 101890-946
4-{2-[(3-Bromobenzyl)oxy]ethyl}piperidine hydrochloride
4-{2-[(3-Bromobenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁BrClNO
MDL Number: MFCD13560662
CAS Number: 1220038-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR