Methyl-4-{[2-(2-piperidinyl)ethoxy]methyl}benzoate hydrochloride

Supplier: Matrix Scientific

050259-500MG 050259-2.5G
101890-936EA 364.62 USD
101890-936 101890-938
Methyl-4-{[2-(2-piperidinyl)ethoxy]methyl}benzoate hydrochloride
Methyl-4-{[2-(2-piperidinyl)ethoxy]methyl}benzoate hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₄ClNO₃
MDL Number: MFCD13560660
CAS Number: 1220038-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR