Methyl-4-{[2-(3-piperidinyl)ethoxy]methyl}benzoate hydrochloride

Supplier: Matrix Scientific

050258-500MG 050258-2.5G
101890-932EA 364.62 USD
101890-932 101890-934
Methyl-4-{[2-(3-piperidinyl)ethoxy]methyl}benzoate hydrochloride
Methyl-4-{[2-(3-piperidinyl)ethoxy]methyl}benzoate hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₄ClNO₃
MDL Number: MFCD13560659
CAS Number: 1220032-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR