Methyl-4-{[2-(4-piperidinyl)ethoxy]methyl}benzoate hydrochloride

Supplier: Matrix Scientific

050257-500MG 050257-2.5G
101890-928EA 364.62 USD
101890-928 101890-930
Methyl-4-{[2-(4-piperidinyl)ethoxy]methyl}benzoate hydrochloride
Methyl-4-{[2-(4-piperidinyl)ethoxy]methyl}benzoate hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₄ClNO₃
MDL Number: MFCD13560658
CAS Number: 1219983-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR