Methyl-2-{[2-(2-piperidinyl)ethoxy]methyl}benzoate hydrochloride

Supplier: Matrix Scientific

050255-500MG 050255-2.5G
101890-920EA 364.62 USD
101890-920 101890-922
Methyl-2-{[2-(2-piperidinyl)ethoxy]methyl}benzoate hydrochloride
Methyl-2-{[2-(2-piperidinyl)ethoxy]methyl}benzoate hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₄ClNO₃
MDL Number: MFCD13560656
CAS Number: 1219964-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR