Methyl-2-{[2-(3-piperidinyl)ethoxy]methyl}benzoate hydrochloride

Supplier: Matrix Scientific

050254-500MG 050254-2.5G
101890-916EA 364.62 USD
101890-916 101890-918
Methyl-2-{[2-(3-piperidinyl)ethoxy]methyl}benzoate hydrochloride
Methyl-2-{[2-(3-piperidinyl)ethoxy]methyl}benzoate hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₄ClNO₃
MDL Number: MFCD13560655
CAS Number: 1220030-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR