Methyl-2-{[2-(4-piperidinyl)ethoxy]methyl}benzoate hydrochloride

Supplier: Matrix Scientific

050253-500MG 050253-2.5G
101890-912EA 364.62 USD
101890-912 101890-914
Methyl-2-{[2-(4-piperidinyl)ethoxy]methyl}benzoate hydrochloride
Methyl-2-{[2-(4-piperidinyl)ethoxy]methyl}benzoate hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₄ClNO₃
MDL Number: MFCD13560654
CAS Number: 1220017-49-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR