3-[(4-Bromo-2-methylphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050252-500MG 050252-2.5G
101890-908EA 364.62 USD
101890-908 101890-910
3-[(4-Bromo-2-methylphenoxy)methyl]pyrrolidine hydrochloride
3-[(4-Bromo-2-methylphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇BrClNO
MDL Number: MFCD13560653
CAS Number: 1220038-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR