2-[2-(4-Bromo-2-methylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050251-500MG 050251-2.5G
101890-904EA 364.62 USD
101890-904 101890-906
2-[2-(4-Bromo-2-methylphenoxy)ethyl]piperidine hydrochloride
2-[2-(4-Bromo-2-methylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁BrClNO
MDL Number: MFCD13560652
CAS Number: 1220030-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR