3-[2-(4-Bromo-2-methylphenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
050250-500MG
050250-2.5G
101890-900EA
364.62
USD
101890-900
101890-902
3-[2-(4-Bromo-2-methylphenoxy)ethyl]piperidine hydrochloride
3-[2-(4-Bromo-2-methylphenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₁BrClNO |
MDL Number:
MFCD13560651 CAS Number: 1220016-63-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...