4-[2-(4-Bromo-2-methylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050249-500MG 050249-2.5G
101890-896EA 364.62 USD
101890-896 101890-898
4-[2-(4-Bromo-2-methylphenoxy)ethyl]piperidine hydrochloride
4-[2-(4-Bromo-2-methylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁BrClNO
MDL Number: MFCD13560650
CAS Number: 1220016-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR