3-[(2-Methoxy-4-propylphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050248-500MG 050248-2.5G
101890-892EA 364.62 USD
101890-892 101890-894
3-[(2-Methoxy-4-propylphenoxy)methyl]pyrrolidine hydrochloride
3-[(2-Methoxy-4-propylphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO₂
MDL Number: MFCD13560649
CAS Number: 1219964-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR