2-[2-(2-Methoxy-4-propylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050247-500MG 050247-2.5G
101890-888EA 364.62 USD
101890-888 101890-890
2-[2-(2-Methoxy-4-propylphenoxy)ethyl]piperidine hydrochloride
2-[2-(2-Methoxy-4-propylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₈ClNO₂
MDL Number: MFCD13560648
CAS Number: 1219982-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR