4-[2-(2-Methoxy-4-propylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050245-500MG 050245-2.5G
101890-880EA 364.62 USD
101890-880 101890-882
4-[2-(2-Methoxy-4-propylphenoxy)ethyl]piperidine hydrochloride
4-[2-(2-Methoxy-4-propylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₈ClNO₂
MDL Number: MFCD13560646
CAS Number: 1220030-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR