3-[2-(2-Benzyl-4-chlorophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050242-500MG 050242-2.5G
101890-868EA 364.62 USD
101890-868 101890-870
3-[2-(2-Benzyl-4-chlorophenoxy)ethyl]piperidine hydrochloride
3-[2-(2-Benzyl-4-chlorophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₂₀H₂₅Cl₂NO
MDL Number: MFCD13560643
CAS Number: 1219982-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR