3-[(2,4-Dichloro-3,5-dimethylphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050240-500MG 050240-2.5G
101890-860EA 364.62 USD
101890-860 101890-862
3-[(2,4-Dichloro-3,5-dimethylphenoxy)methyl]pyrrolidine hydrochloride
3-[(2,4-Dichloro-3,5-dimethylphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈Cl₃NO
MDL Number: MFCD13560641
CAS Number: 1220032-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR