3-[2-(2,4-Dichloro-3,5-dimethylphenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
050238-500MG
050238-2.5G
101890-852EA
364.62
USD
101890-852
101890-854
3-[2-(2,4-Dichloro-3,5-dimethylphenoxy)ethyl]piperidine hydrochloride
3-[2-(2,4-Dichloro-3,5-dimethylphenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₂Cl₃NO |
MDL Number:
MFCD13560639 CAS Number: 1220016-59-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...