4-[2-(2,4-Dichloro-3,5-dimethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050237-500MG 050237-2.5G
101890-848EA 364.62 USD
101890-848 101890-850
4-[2-(2,4-Dichloro-3,5-dimethylphenoxy)ethyl]piperidine hydrochloride
4-[2-(2,4-Dichloro-3,5-dimethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₂Cl₃NO
MDL Number: MFCD13560638
CAS Number: 1219972-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR