Benzyl-4-(3-pyrrolidinylmethoxy)benzoate hydrochloride

Supplier: Matrix Scientific

050236-500MG 050236-2.5G
101890-844EA 364.62 USD
101890-844 101890-846
Benzyl-4-(3-pyrrolidinylmethoxy)benzoate hydrochloride
Benzyl-4-(3-pyrrolidinylmethoxy)benzoate hydrochloride

Hazard: Irritant

Formula: C₁₉H₂₂ClNO₃
MDL Number: MFCD13560637
CAS Number: 1220032-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR