Benzyl-4-[2-(3-piperidinyl)ethoxy]benzoate hydrochloride
Supplier: Matrix Scientific
050234-500MG
050234-2.5G
101890-836EA
364.62
USD
101890-836
101890-838
Benzyl-4-[2-(3-piperidinyl)ethoxy]benzoate hydrochloride
Benzyl-4-[2-(3-piperidinyl)ethoxy]benzoate hydrochloride
Hazard: Irritant
Formula:
C₂₁H₂₆ClNO₃ MW: 375.9 g/mol |
MDL Number:
MFCD13560635 CAS Number: 1220016-50-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...