Benzyl-4-[2-(4-piperidinyl)ethoxy]benzoate hydrochloride

Supplier: Matrix Scientific

050233-500MG 050233-2.5G
101890-832EA 364.62 USD
101890-832 101890-834
Benzyl-4-[2-(4-piperidinyl)ethoxy]benzoate hydrochloride
Benzyl-4-[2-(4-piperidinyl)ethoxy]benzoate hydrochloride

Hazard: Irritant

Formula: C₂₁H₂₆ClNO₃
MW: 375.9 g/mol
MDL Number: MFCD13560634
CAS Number: 906744-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR