3-[(3-Ethoxyphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050232-500MG 050232-2.5G
101890-828EA 364.62 USD
101890-828 101890-830
3-[(3-Ethoxyphenoxy)methyl]pyrrolidine hydrochloride
3-[(3-Ethoxyphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO₂
MDL Number: MFCD13560633
CAS Number: 1220016-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR