2-[2-(3-Ethoxyphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050231-500MG 050231-2.5G
101890-824EA 364.62 USD
101890-824 101890-826
2-[2-(3-Ethoxyphenoxy)ethyl]piperidine hydrochloride
2-[2-(3-Ethoxyphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO₂
MDL Number: MFCD13560632
CAS Number: 1220017-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR