3-[2-(3-Ethoxyphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050230-500MG 050230-2.5G
101890-820EA 364.62 USD
101890-820 101890-822
3-[2-(3-Ethoxyphenoxy)ethyl]piperidine hydrochloride
3-[2-(3-Ethoxyphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO₂
MDL Number: MFCD13560631
CAS Number: 1219982-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR