4-[2-(3-Ethoxyphenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
050229-500MG
050229-2.5G
101890-816EA
364.62
USD
101890-816
101890-818
4-[2-(3-Ethoxyphenoxy)ethyl]piperidine hydrochloride
4-[2-(3-Ethoxyphenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₄ClNO₂ |
MDL Number:
MFCD13560630 CAS Number: 1219982-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...