3-[(2-Iodophenoxy)methyl]pyrrolidine hydrochloride
Supplier: Matrix Scientific
050228-500MG
050228-2.5G
101890-812EA
364.62
USD
101890-812
101890-814
3-[(2-Iodophenoxy)methyl]pyrrolidine hydrochloride
3-[(2-Iodophenoxy)methyl]pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₅ClINO |
MDL Number:
MFCD13560629 CAS Number: 1220032-37-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...