3-[2-(2-Iodophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050226-500MG 050226-2.5G
101890-804EA 364.62 USD
101890-804 101890-806
3-[2-(2-Iodophenoxy)ethyl]piperidine hydrochloride
3-[2-(2-Iodophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClINO
MDL Number: MFCD13560627
CAS Number: 1219972-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR