3-[(4-Bromo-2-fluorophenoxy)methyl]pyrrolidine hydrochloride
Supplier: Matrix Scientific
050224-500MG
050224-2.5G
101890-796EA
364.62
USD
101890-796
101890-798
3-[(4-Bromo-2-fluorophenoxy)methyl]pyrrolidine hydrochloride
3-[(4-Bromo-2-fluorophenoxy)methyl]pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₄BrClFNO |
MDL Number:
MFCD13560625 CAS Number: 1219982-63-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...