4-[2-(4-Bromo-2-fluorophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050221-500MG 050221-2.5G
101890-784EA 364.62 USD
101890-784 101890-786
4-[2-(4-Bromo-2-fluorophenoxy)ethyl]piperidine hydrochloride
4-[2-(4-Bromo-2-fluorophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈BrClFNO
MDL Number: MFCD13560622
CAS Number: 1220016-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR