3-[(2-Nitrophenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050220-500MG 050220-2.5G
101890-780EA 364.62 USD
101890-780 101890-782
3-[(2-Nitrophenoxy)methyl]pyrrolidine hydrochloride
3-[(2-Nitrophenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅ClN₂O₃
MDL Number: MFCD13560621
CAS Number: 1220016-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR