2-[2-(2-Nitrophenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
050219-500MG
050219-2.5G
101890-776EA
364.62
USD
101890-776
101890-778
2-[2-(2-Nitrophenoxy)ethyl]piperidine hydrochloride
2-[2-(2-Nitrophenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₉ClN₂O₃ |
MDL Number:
MFCD13560620 CAS Number: 1219982-53-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...