3-[2-(2-Nitrophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050218-500MG 050218-2.5G
101890-772EA 364.62 USD
101890-772 101890-774
3-[2-(2-Nitrophenoxy)ethyl]piperidine hydrochloride
3-[2-(2-Nitrophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O₃
MDL Number: MFCD13560619
CAS Number: 1220032-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR