4-[2-(2-Nitrophenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
050217-500MG
050217-2.5G
101890-768EA
364.62
USD
101890-768
101890-770
4-[2-(2-Nitrophenoxy)ethyl]piperidine hydrochloride
4-[2-(2-Nitrophenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₉ClN₂O₃ |
MDL Number:
MFCD13560618 CAS Number: 1219982-52-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...