3-[(4-Isopropyl-3-methylphenoxy)methyl]pyrrolidine hydrochloride
Supplier: Matrix Scientific
050216-500MG
050216-2.5G
101890-764EA
364.62
USD
101890-764
101890-766
3-[(4-Isopropyl-3-methylphenoxy)methyl]pyrrolidine hydrochloride
3-[(4-Isopropyl-3-methylphenoxy)methyl]pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₄ClNO |
MDL Number:
MFCD13560617 CAS Number: 1220032-36-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...