2-[2-(4-Isopropyl-3-methylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050215-500MG 050215-2.5G
101890-760EA 364.62 USD
101890-760 101890-762
2-[2-(4-Isopropyl-3-methylphenoxy)ethyl]piperidine hydrochloride
2-[2-(4-Isopropyl-3-methylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₈ClNO
MDL Number: MFCD13560616
CAS Number: 1220032-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR