3-[2-(4-Isopropyl-3-methylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050214-500MG 050214-2.5G
101890-756EA 364.62 USD
101890-756 101890-758
3-[2-(4-Isopropyl-3-methylphenoxy)ethyl]piperidine hydrochloride
3-[2-(4-Isopropyl-3-methylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₈ClNO
MDL Number: MFCD13560615
CAS Number: 1220032-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR