4-[2-(4-Isopropyl-3-methylphenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
050213-500MG
050213-2.5G
101890-752EA
364.62
USD
101890-752
101890-754
4-[2-(4-Isopropyl-3-methylphenoxy)ethyl]piperidine hydrochloride
4-[2-(4-Isopropyl-3-methylphenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₇H₂₈ClNO |
MDL Number:
MFCD13560614 CAS Number: 1220017-13-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...