3-[(2-Bromo-4-fluorophenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050212-500MG 050212-2.5G
101890-748EA 364.62 USD
101890-748 101890-750
3-[(2-Bromo-4-fluorophenoxy)methyl]pyrrolidine hydrochloride
3-[(2-Bromo-4-fluorophenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄BrClFNO
MW: 310.6 g/mol
MDL Number: MFCD13560613
CAS Number: 1219964-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR