2-[2-(2-Bromo-4-fluorophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050211-500MG 050211-2.5G
101890-744EA 364.62 USD
101890-744 101890-746
2-[2-(2-Bromo-4-fluorophenoxy)ethyl]piperidine hydrochloride
2-[2-(2-Bromo-4-fluorophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈BrClFNO
MDL Number: MFCD13560612
CAS Number: 1219964-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR